Learn More
Dibutyl Disulfide, Sigma-Aldrich™
Product Code. 15894501
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894501 25 mL 25mL
15804511 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894501 Supplier Sigma Aldrich Supplier No. 8202420025
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 629-45-8
Specifications
Melting Point -94°C
Density 0.94 g/cm3 at 20°C
Molecular Formula CH3(CH2)3SS(CH2)3CH3
MDL Number MFCD00009467
Quantity 25 mL
Synonym Dibutyl disulfide, Butyl disulfide, Dibutyl disulfide
Molecular Weight (g/mol) 178.36
Grade Research