Learn More
Dibrompropamidine Diisetionate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Dibrompropamidine diisetionate |
| CAS | 614-87-9 |
| Analyte or Component Names | Dibrompropamidine diisetionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 Br2 N4 O2 . 2 C2 H6 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-(trimethylenedioxy)bis[3-bromobenzamidine], Benzamidine, 4,4'-(trimethylenedioxy)bis[3-bromo-, bis(2-hydroxyethanesulfonate), Benzenecarboximidamide, 4,4'-[1,3-propanediylbis(oxy)]bis[3-bromo-, bis(2-hydroxyethanesulfonate), 1,3-Di(4-amidino-2-bromophenoxy)propane di-2-hydroxyethanesulfonate, 2,2'-Dibromo-4,4'-diamidinodiphenoxypropane di-(β-hydroxyethanesulfonate), Brolene, Brolene Ointment, Brulidine, Dibromopropamidine Diisethionate, Dibromopropamidine isethionate, Dibrompropamidine isethionate, NSC 33424, NSC 528031 |
| InChI Formula | InChI=1S/C17H18Br2N4O2.2C2H6O4S/c18-12-8-10(16(20)21)2-4-14(12)24-6-1-7-25-15-5-3-11(17(22)23)9-13(15)19;2*3-1-2-7(4,5)6/h2-5,8-9H,1,6-7H2,(H3,20,21)(H3,22,23);2*3H,1-2H2,(H,4,5,6) |
| Solution Type | Neat |
| SMILES | NC(=N)c1ccc(OCCCOc2ccc(cc2Br)C(=N)N)c(Br)c1.OCCS(=O)(=O)O.OCCS(=O)(=O)O |
| Show More |