Learn More
Dibenzothiophene 5-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Dibenzothiophene 5-Oxide |
| CAS | 1013-23-6 |
| Molecular Formula | C12H8OS |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C12H8OS/c13-14-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8H |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3S2=O |
| Molecular Weight (g/mol) | 200.25 |
| Formula Weight | 200.0296 |