Learn More
Dibenzofuran, Sigma-Aldrich™
Product Code. 15860941
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860941 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15860941 Supplier Sigma Aldrich Supplier No. 8204080250
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 132-64-9
Specifications
Melting Point 80°C to 82°C (External MSDS)
Density 1.3 g/cm3 at 20°C
MDL Number MFCD00004968
Quantity 250 g
Synonym Dibenzofuran, Diphenylene oxide, Dibenzofuran
Molecular Weight (g/mol) 168.19
Grade Research