Learn More
Dibenzo[b,f][1,4]thiazepin-11(10H)-one, Mikromol™
Description
Dibenzo[b,f][1,4]thiazepin-11(10H)-one
Specifications
Specifications
| Chemical Name or Material | Dibenzo[b,f][1,4]thiazepin-11(10H)-one |
| CAS | 7-7-3159 |
| Analyte or Component Names | Dibenzo[b,f][1,4]thiazepin-11(10H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H9 N O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dibenzo[b,f][1,4]thiazepin-11(10H)-one, Quetiapine Fumarate Imp. G (EP), Quetiapine Related Compound G, Quetiapine Fumarate Impurity G, Quetiapine Impurity G |
| InChI Formula | InChI=1S/C13H9NOS/c15-13-9-5-1-3-7-11(9)16-12-8-4-2-6-10(12)14-13/h1-8H,(H,14,15) |
| SMILES | O=C1Nc2ccccc2Sc3ccccc13 |
| IUPAC Name | 5H-benzo[b][1,4]benzothiazepin-6-one |
| Show More |
Product Title
¿Detecta una oportunidad de mejora?