Learn More
Diatomaceous earth, Calcined, Honeywell™ Fluka™
Product Code. p-7120368 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
5kg
Product Code. 15677810
Brand: Honeywell-Fluka D3877500G
Request Bulk or Custom Format
Powder, suitable for most filtrations
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]
Specifications
Quantity 500 g
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Diatomaceous Earth, Calcined