Learn More
Macherey-Nagel™ DiaQuant™ Peracetic Acid
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
100 Test Strips/Pk.
Unit Size:
Each
Description
- Specific to peracetic acid and it will not detect peroxides.
- The easy Dip and Read procedure provides reliable results in 10 seconds.
- Technicians may clean hemodialysis equipment using peracetic acid containing reagents (for example, Puristeril plus).
- Prior to the next use, they must rinse the system, until the reagent is completely washed out.
- Provides a quick, easy and dependable method to check rinse water and ensure safe dialysis.
Specifications
Specifications
| Color | White to dark blue color change |
| Packaging | 100 Test Strips/Pk. |
| Quantity | 100 Test Strips/Pk. |
| Range | 0, 5, 10, 20, 30, 50 mg/L PAA |
| Type | Colorimetric, semiquantitative test strip |