Learn More
Checkers Diamondback™ Ramp
Product Code. 17702508
Change view
Click to view available options
Color:
Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17702508 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17702508 Supplier Checkers Supplier No. PDBS0002YL
  • ONLINE EXCLUSIVE PRICE
    4143.12 NOK
    3373.20 NOK / Pack of 1
    Save 769.92 NOK
    18% Off
Estimated Shipment: 21-12-2026
Diamondback 4.5 in. hose bridge system provides protection for valuable cable and hose lines while ensuring a safer method of passage for vehicles. They can protect hoses and cables up to 4.5 in.
  • Checkers Diamondback 45 in. hose bridge system provides protection for valuable cable and hose lines while ensuring a safer method of passage for vehicles
  • They replace wooden and metal fabricated ramps and can be used to protect lines up to 45 inches in diameter
  • High density, all-weather polyurethane construction protects from damage by physical pressure and reduces cable and hose maintenance and replacement costs
  • The lightweight, modular design of interlocking sections provides multi-length flexibility
  • Checkers Hose Bridge Systems are intended for temporary installations only
Specifications
Dimensions (L x W x H) 41 x 26 x 14 cm
Color Yellow
Material Polyethylene
Type Ramp