Learn More
Scotch-Brite™ Diamond Floor Pads, Purple
Description
- Simultaneously cleans and polishes flooring to achieve an outstanding shine
- Ideal for marble, terrazzo, vinyl, slate, and polished concrete floors
- Easy to use: no expensive or harmful chemicals needed, only water
- Recommended for use with low and standard speed machines
Specifications
Specifications
| Product Type | Cleaning Pads |
| Color | Purple |
| Diameter (Metric) | 355 mm |
| For Use With (Application) | Burnishing, Cleaning |
| For Use With (Equipment) | Marble, Terrazzo, Slate, and Polished Concrete Flooring |
| Material | Diamond-coated |
| Style | Round |