Learn More
Diaion™ WK60L(H), ion exchange resin, weakly acidic porous type, 4.4 meq/ml on polyacrylate, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046797.A1
Additional Details : Weight : 1.00000kg
Descrizione
Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications. Can be used in wide pH range
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications
- Can be used in wide pH range
Specifica
425-1180μm Beads | |
Weakly acidic porous type, 4.4meq/mL on polyacrylate | |
1 kg |
Diaion™ WK40L (H), ion exchange resin | |
425 to 1180μm |
RUO – Research Use Only