Learn More
MP Biomedicals Diagnostic Kit for Triiodothyronine (T3) (MPQuanti™ only)
Product Code. 30280606 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Total T3
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280606 Total T3 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280606 Supplier MP Biomedicals Supplier No. 07T35025
  • ONLINE EXCLUSIVE PRICE
    976.99 NOK
    921.48 NOK / 1 sets
    Save 55.51 NOK
    5% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Diagnostic Kit for Triiodothyronine (T3) is intended for in-vitro quantitative detection of total Triiodothyronine (T3) in human serum or plasma.
  • Diagnostic Kit for Triiodothyronine (T3) (Serum/Plasma) is based on fluorescence immunoassay technology.
  • The specimen moves through the strip from sample pad to absorbent pad. T3 in the specimen will compete with the T3 antigen coated on the strip.
  • The less T3 in the specimen, the more fluorescent microspheres conjugated with anti-T3 antibodies can be captured by the T3 antigen coated on the strip.
  • The concentration of T3 in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of T3 in the sample is calculated by the, compatible only with the MPQuanti™ Immuflorescence Analyzer, to show T3 concentration in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 62 to 9.24nmol/L, R≥0.990
  • Test results should be interpreted at 15 minutes
Specifications
Detectable Analytes Total T3
Detection Method Immunofluorescence
Sample Type Human serum, Human plasma
Type POC, Quantitative, T3, Triiodothyronine
Form Ready-to-Use (RTU)