Learn More
MP Biomedicals Diagnostic Kit for Progesterone (MPQuanti™ only)
Product Code. 30280621 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Progesterone
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280621 Progesterone 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280621 Supplier MP Biomedicals Supplier No. 07PROG5025
  • ONLINE EXCLUSIVE PRICE
    1221.11 NOK
    1151.72 NOK / 1 sets
    Save 69.39 NOK
    5% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Diagnostic Kit for Progesterone is intended for in-vitro quantitative determination of Progesterone in human serum, and plasma.
  • Diagnostic Kit for Progesterone (Serum/Plasma) is based on fluorescence immunoassay technology.
  • The specimen moves through the strip from sample pad to absorbent pad. Progesterone in the specimen will compete with the Progesterone antigen coated on the membrane with the progesterone antibody labeled with fluorescent microspheres.
  • The less Progesterone in the specimen, the more chance that fluorescent microspheres-conjugated anti-Progesterone antibodies can be captured by the Progesterone antigen coated on the membrane (Test line).
  • The concentration of Progesterone in the specimen is inversely related to the intensity of the fluorescent signal captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of Progesterone in the specimen is calculated by the MPQuanti™ Immuflorescence Analyzer to show Progesterone concentration in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1.4 ̴ 60 ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
Specifications
Detectable Analytes Progesterone
Detection Method Immunofluorescence
Sample Type Human serum, Human plasma
Type POC, Quantitative, progesterone, hormone
Form Ready-to-Use (RTU)