Learn More
MP Biomedicals Diagnostic Kit for D-Dimer (MPQuanti™ only)
Product Code. 30280622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280622 25 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280622 Supplier MP Biomedicals Supplier No. 07CTNL5025
  • ONLINE EXCLUSIVE PRICE
    814.12 NOK
    767.88 NOK / 1 sets
    Save 46.24 NOK
    5% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Diagnostic Kit for D-Dimer is intended for in-vitro quantitative detection of D-Dimer in human whole blood or plasma.
  • Diagnostic Kit for D-Dimer (Whole Blood/Plasma) is based on fluorescence immunoassay technology to measure D-Dimer in human whole blood or plasma as an aid in the diagnosis of DVT and PE.
  • The test is intended for healthcare professionals use.
  • The sample moves through the strip from sample pad to absorbent pad. If the specimen contains D-Dimer, it attaches to the fluorescent microspheres-conjugated anti-D-Dimer antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of D-Dimer in the sample correlates with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of D-Dimer in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1 ̴10 mg/L, R≥0.990
  • Test results should be interpreted at 15 minutes
  • Used as an aid in the diagnosis of DVT and PE
Specifications
Detectable Analytes D-dimer
Sample Type Human whole blood, Human plasma
Type POC, Quantitative, D-Dimer
Quantity 25 Tests/Kit
Form Ready-to-Use (RTU)