Learn More
MP Biomedicals Diagnostic Kit for CK-MB (MPQuanti™ only)
Product Code. 30280615 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280615 25 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280615 Supplier MP Biomedicals Supplier No. 07CKMB5025
  • ONLINE EXCLUSIVE PRICE
    814.12 NOK
    767.88 NOK / 1 sets
    Save 46.24 NOK
    5% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Diagnostic Kit for CK-MB is a chromatographic immunoassay for the quantitative detection of human CK-MB in human whole blood, serum or plasma.
  • The CK-MB Test Cassette (Whole Blood/Serum/Plasma) is a chromatographic immunoassay for the quantitative detection of human CK-MB in human whole blood, serum or plasma as an aid in the diagnosis of myocardial infarction (MI).
  • The sample moves through the strip from sample pad to absorbent pad.
  • If the test sample contains CK-MB, it attaches to the CK-MB antibody which is conjugated with fluorescent microspheres.
  • Then the complex will be captured by the capture antibody coated on the nitrocellulose membrane (Test line).
  • The concentration of CK-MB in the sample correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and product standard curve, the concentration of CK-MB in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE-marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 0. 2 ̴75ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
Specifications
Detectable Analytes CK-MB
Detection Method Immunofluorescence
Sample Type Human whole blood, Human serum, Human plasma
Type POC, CK-MB, Quantitative
Quantity 25 Tests/Kit
Form Ready-to-Use (RTU)