Learn More
MP Biomedicals Diagnostic Kit for Cardiac Troponin I (MPQuanti™ only)
Product Code. 30280598 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280598 25 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280598 Supplier MP Biomedicals Supplier No. 07CTN15025
  • ONLINE EXCLUSIVE PRICE
    651.26 NOK
    614.27 NOK / 1 sets
    Save 36.99 NOK
    5% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Diagnostic Kit for Cardiac Troponin I is intended for in-vitro quantitative determination of human cardiac Troponin I (cTnI) in whole blood, serum, or plasma.
  • Diagnostic Kit for Cardiac Troponin I (Whole Blood/Serum/Plasma) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad.
  • If the specimen contains cTnI, it attaches to the fluorescent microspheres-conjugated anti-cTnI antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of cTnI in the sample correlates with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and product standard curve, the concentration of cTnI in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in vitro diagnostic use
  • Working with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1 ̴40 ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
  • Used as an aid in diagnosis of Myocardial Infarction (MI)
Specifications
Detectable Analytes Troponin I
Sample Type Human whole blood, Human serum, Human plasma
Type POC, cTnI, Quantitative
Quantity 25 Tests/Kit
Form Ready-to-Use (RTU)