missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Dicarbonate, Sigma-Aldrich™

Product Code. 15870331
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870331 25 mL 25mL
15816570 100 mL 100mL
15826570 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15870331 Supplier Sigma Aldrich Supplier No. 8182820025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24424-99-5

Specifications

Melting Point 20°C to 24°C
Density 1.02 g/cm3 at 20°C
Boiling Point 56°C to 57°C/0.7 hPa
MDL Number MFCD00008805
Quantity 25 mL
Synonym Di-tert-butyl dicarbonate, Di-tert-butyl dicarbonate
Molecular Weight (g/mol) 218.25
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.