Learn More
di-Sodium Tetraborate Decahydrate, Supelco™
Product Code. 15894790
Change view
Click to view available options
Quantity:
500 g
1 kg
50 kg
Unit Size:
1kg
500g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894790 1 kg 1kg
15884790 500 g 500g
15887440 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15894790 Supplier Supelco Supplier No. 1063081000
 Restricted Item
Estimated Shipment: 12-11-2026
For analysis ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1303-96-4
Specifications
Melting Point 75°C (Elimination of water of crystallisation)
Quantity 1 kg
MDL Number MFCD00149193
Synonym di-Sodium tetraborate decahydrate, Borax, Sodium biborate decahydrate, Sodium borate decahydrate, di-Sodium tetraborate decahydrate
Molecular Weight (g/mol) 381.37
Grade Research
Density 1.72 g/cm3 at 20°C
Chemical Name or Material Di-Sodium Tetraborate Decahydrate