missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Tetraborate Decahydrate, 99+%, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
3 kg
Unit Size:
1kg
3kg
Specifications
Specifications
| Melting Point | 75°C |
| Color | White |
| pH | 9 |
| Physical Form | Powder/Solid |
| Quantity | 3 kg |
| Formula Weight | 381.36g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium tetraborate decahydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction