missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Oxalate, Supelco™

Product Code. 15894240
Change view
Click to view available options
Quantity:
60 g
Unit Size:
60g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894240 60 g 60g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894240 Supplier Supelco Supplier No. 1024070060
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric standard, secondary reference material for redox titration, traceable to NIST Standard Reference Material (SRM) Certipur™ Reag. USP

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 250°C to 270°C (Decomposition)
CAS 62-76-0
Quantity 60 g
MDL Number MFCD00012465
Synonym di-Sodium oxalate, Oxalic acid sodium salt, di-Sodium oxalate
Molecular Weight (g/mol) 134
Grade Research
Density 2.27 g/cm3 at 20°C
Chemical Name or Material Di-Sodium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.