missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Oxalate, Supelco™

Product Code. 15857490
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15857490 1 kg 1kg
15844860 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15857490 Supplier Supelco Supplier No. 1065571000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-76-0

Specifications

Melting Point 250°C to 270°C (decomposition)
Quantity 1 kg
MDL Number MFCD00012465
Synonym di-Sodium oxalate, Oxalic acid sodium salt, di-Sodium oxalate
Molecular Weight (g/mol) 134
Grade Research
Density 2.27 g/cm3 at 20°C
Chemical Name or Material Di-Sodium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.