Learn More
di-Potassium Phthalate, Sigma-Aldrich™
Description
for synthesis
Specifications
Specifications
| CAS | 4409-98-7 |
| Molecular Formula | C6H4-1,2-(COOK)2 |
| MDL Number | MFCD00040595 |
| Quantity | 100 g |
| Synonym | di-Potassium phthalate, di-Potassium phthalate |
| Molecular Weight (g/mol) | 242.31 |
| Grade | Research |