missing translation for 'onlineSavingsMsg'
Learn More
Learn More
di-Potassium Oxalate Monohydrate, Supelco™
Specifications
Specifications
| Melting Point | >138°C (decomposition) |
| Quantity | 1 kg |
| MDL Number | MFCD00150033 |
| Synonym | di-Potassium oxalate monohydrate, Oxalic acid dipotassium salt monohydrate, di-Potassium oxalate monohydrate |
| Molecular Weight (g/mol) | 166.22 (anhydrous basis) |
| Grade | Research |
| Density | 2.20 g/cm3 at 20.4°C |
| Chemical Name or Material | Di-Potassium Oxalate Monohydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?