missing translation for 'onlineSavingsMsg'
Learn More

Di-p-toluoyl-D-tartaric acid monohydrate, 98%, Thermo Scientific Chemicals

Product Code. p-7077164 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15750809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15750809 Supplier Thermo Scientific Acros Supplier No. 148460250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71607-31-3
Molecular Formula C20H20O9
Molecular Weight (g/mol) 404.37
MDL Number MFCD00149567,MFCD00064440,MFCD00151034
InChI Key FOTRUJUPLHRVNU-UHFFFAOYNA-N
Synonym +-o,o'-di-p-toluoyl-d-tartaric acid monohydrate, 2r,3s-2,3-bis 4-methylbenzoyloxy butanedioic acid hydrate, 2s,3s-+-2,3-bis 4-methylbenzoyl oxy succinic acid monohydrate
PubChem CID 71299557
SMILES O.CC1=CC=C(C=C1)C(=O)OC(C(OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 163.0°C to 165.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula [CH3C6H4CO2CH(CO2H)-]2·H2O
Quantity 25 g
Fieser 01,351
Specific Rotation + 140.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and acetone
IUPAC Name (2R,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
Formula Weight 404.38
Percent Purity 98%
Chemical Name or Material Di-p-toluoyl-D-tartaric acid monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.