Learn More
di-Lithium Oxalate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 553-91-3 |
| MDL Number | MFCD00040596 |
| Quantity | 100 g |
| Synonym | di-Lithium oxalate, Oxalic acid dilithium salt, di-Lithium oxalate |
| Molecular Weight (g/mol) | 101.9 |
| Grade | Research |