Learn More
Di(ethylene glycol) divinyl ether, 95%
Product Code. 15274714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15274714 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15274714 Supplier Thermo Scientific Acros Supplier No. 395410500
  • ONLINE EXCLUSIVE PRICE
    1960.00 NOK
    1710.65 NOK / 50mL
    Save 249.35 NOK
    12% Off
 Restricted Item
In Stock
Add to basket
CAS: 764-99-8 | C8H14O3 | 158.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 764-99-8
Melting Point -21.0°C
Density 0.9700g/mL
Boiling Point 168.0°C
Flash Point 88°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C8H14O3
Linear Formula CH2CHOCH2CH2OCH2CH2OCHCH2
Quantity 50 mL
Specific Gravity 0.97
Synonym diethylene glycol divinyl ether, divinylcarbitol, dvedeg, dvedeg russian, ether, bis 2-vinyloxy ethyl, ethene, 1,1'-oxybis 2,1-ethanediyloxy bis, 3,6,9-trioxaundeca-1,10-diene, unii-abm90y2g95, divinyl ether diethylenglykolu, bis 2-vinyloxy ethyl ether
Solubility Information Solubility in water: insoluble.
InChI Key SAMJGBVVQUEMGC-UHFFFAOYSA-N
SMILES C=COCCOCCOC=C
IUPAC Name 1-ethenoxy-2-(2-ethenoxyethoxy)ethane
Molecular Weight (g/mol) 158.19
PubChem CID 12998
Formula Weight 158.19
Percent Purity 95%
Chemical Name or Material Di(ethylene glycol) divinyl ether