Learn More
di-Ammonium Hydrogen Citrate, Supelco™
Product Code. 15894030
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894030 500 g 500g
15826960 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894030 Supplier Supelco Supplier No. 1011540500
 Restricted Item
Estimated Shipment: 18-11-2026
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3012-65-5
Specifications
Quantity 500 g
MDL Number MFCD00013068
Synonym di-Ammonium hydrogen citrate, Ammonium citrate dibasic, Diammonium hydrogen citrate, di-Ammonium hydrogen citrate
Molecular Weight (g/mol) 226.18
Grade Research
Density 1.48 g/cm3 at 20°C
Chemical Name or Material Di-Ammonium Hydrogen Citrate