Learn More
Di-2-ethylhexyl Phthalate, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 117-81-7 |
| Molecular Formula | C24H38O4 |
| Quantity | 100 g |
| Synonym | dehp, bis 2-ethylhexyl phthalate, di 2-ethylhexyl phthalate, diethylhexyl phthalate, di-sec-octyl phthalate, octyl phthalate, fleximel, octoil |
| InChI Key | BJQHLKABXJIVAM-UHFFFAOYSA-N |
| SMILES | CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC |
| IUPAC Name | bis(2-ethylhexyl) benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 390.564 |
| PubChem CID | 8343 |
| ChEBI | CHEBI:17747 |
| Show More |