missing translation for 'onlineSavingsMsg'
Learn More

DHX9 Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 1B12C10, Proteintech

Product Code. 17701309 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17701309 20 μL 20µL
17649876 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17701309

Brand: Proteintech CL4886715320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DHX9 Monoclonal antibody apecifically detetects DHX9 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence, Flow Cytometry.

Specifications

Antigen DHX9
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 1B12C10
Concentration 1000 ug/mL
Conjugate CoraLite 488
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC014246
Gene Symbols DHX9
Host Species Mouse
Immunogen DHX9 fusion protein Ag12104
Purification Method Protein G purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1660
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.