Learn More
dGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16459-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16886084
  • 1611.84 NOK / 20µL
Estimated Shipment: 13-08-2024
to see stock.
Description
Description
Mitochondrial deoxyguanosine kinase (DGUOK) is required for the phosphorylation of several deoxyribonucleosides and certain purine deoxykribonucleoside analogs widely employed as antiviral and chemotherapeutic agents. Purine deoxyribonucleoside analogs are extensively used in treatment of lymphoproliferative disorders. These compounds are administered as pro-drugs, and their efficiency is dependent on intracellular phosphorylation to the corresponding triphosphates. In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predomitly by 2 deoxyribonucleoside kinases: cytosolic deoxycytidine kinase (DCK) and mitochondrial deoxyguanosine kinase (DGUOK also known as DGK). DGUOK expression is ubiquitous, with highest levels in muscle, brain, liver and lymphoid tissues. Defects in DGUOK are a cause of mitochondrial DNA depletion syndrome (MDS). MDS is a clinically heterogeneous group of disorders characterized by a reduction in mitochondrial DNA (mtDNA) copy number. Primary mtDNA depletion is inherited as an autosomal recessive trait and may affect single organs, typically muscle or liver, or multiple tissues.
Specifications
Specifications
dGK
Polyclonal
Unconjugated
DGUOK
deoxyguanosine kinase, dGK, DGUOK
Rabbit
Antigen Affinity Chromatography
RUO
1716, 27369, 297389
-20°C
Liquid
Immunoprecipitation
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16854, Q9QX60
DGUOK
dGK Fusion Protein Ag9501
20 μL
Primary
Mouse, Rat, Human