Learn More
Dextran Sulfate Sodium Salt
Product Code. 15895118 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895118 100 g Plastic bottle 100g
15875118 10 g Plastic bottle 10g
15885118 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15895118 Supplier Thermo Scientific Supplier No. J14489.22
  • ONLINE EXCLUSIVE PRICE
    12500.00 NOK
    9387.65 NOK / 100g
    Save 3112.35 NOK
    24% Off
 Restricted Item
In Stock
Add to basket
CAS: 9011-18-1 ∣ (C6H7O5)m(C6H7O5)n ∣ 40,000 g/mol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
Specifications
Physical Form Powder
Quantity 100 g
Packaging Plastic bottle
Formula Weight 40,000- 50,000 Kd
Grade Ungraded
Chemical Name or Material Dextran Sulfate Sodium Salt