missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ DexPure Vinyl Gloves

Product Code. 12616093
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Powdered:
Powdered
Color:
Blue
Translucent
Unit Size:
Pack of 100
Product Code. Size Powdered Color unitSize
12616093 Small Powdered Translucent Pack of 100
12696083 Large Powdered Translucent Pack of 100
12606093 Medium Powdered Translucent Pack of 100
12686103 X-Large Powdered Blue Pack of 100
12626093 X-Large Powdered Translucent Pack of 100
5 options
This item is not returnable. View return policy

Product Code. 12616093

Brand: Honeywell Safety Products™ 4580001S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Convenient and disposable, these translucent vinyl gloves can be used for a variety of tasks from the laboratory to the kitchen. Honeywell Safety Products™ DexPure Vinyl Gloves also have a smooth finish that provides great dexterity.

  • Translucent vinyl glove.
  • Smooth finish for great dexterity.
  • Certified for food contact and treated to prevent bacteria.
  • Suitable for the following:
    • Product inspection.
    • Precision assembly.
    • Laboratory work.
    • Handling of paints, inks and dyes.
  • Single use.
  • AQL:1.5.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive n°1934/2004 : Food contact, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0465, EC Attestation Number 0075/797/162/11/08/0465
Classification Dexterity: 5
Description AQL:1.5
Color Translucent
Disposable Single-use
Material Vinyl
Powdered Powdered
Product Type Gloves
Size Small
Thickness (Metric) 0.1 mm
For Use With (Application) Automotive, Aviation, Catering, Chemical, Food, Industrial Cleaning, Laboratory, Maintenance, Medical, Pharmaceutical, Petrochemical, Printing and Water Treatment industries.
Quantity 100/PK
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.