Learn More
Dexamethasone 9,11-Epoxide, TRC
Product Code. 30009965
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009965 100 mg 100mg
30011519 1 g 1g
30006167 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009965 Supplier Toronto Research Chemicals Supplier No. TRCD298795100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24916-90-3
Molecular Formula C22 H28 O5
Molecular Weight (g/mol) 372.45
Synonym 17,21-Dihydroxy-16α-methyl-9β,11β-epoxypregna-1,4-diene-3,20-dione, 9β,11β-Epoxy-17,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, Dexamethasone 9,11-Epoxide
IUPAC Name 17,21-Dihydroxy-16α-methyl-9β,11β-epoxypregna-1,4-diene-3,20-dione
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CO
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15)18(27-22)10-20(16,3)21(12,26)17(25)11-23/h6-7,9,12,15-16,18,23,26H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,18+,19+,20+,21+,22-/m1/s1
Formula Weight 372.1937
Percent Purity >95%
Chemical Name or Material 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione
Grade HPLC