Learn More
KGW Isotherm™ Dewar Large Insulating Flasks
Product Code. 10382801
Change view
Click to view available options
Capacity (Metric):
7 L
14 L
Height (Metric):
385mm
610mm
Depth (Metric) Inside:
275mm
500mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Depth (Metric) Inside Unit Size
10382801 14 L 610mm 500mm Each
10403501 7 L 385mm 275mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10382801 Supplier KGW Isotherm™ Supplier No. 1243
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
For the storage and transportation of temperature-sensitive dry goods such as dry ice. KGW Isotherm™ Dewar Large Insulating Flasks feature a robust design made from borosilicate glass 3.3.
  • Large aperture
  • Insulated lid (including grip)
  • Fully silvered as standard
  • Quick-release clamp (sold separately)
Specifications
Cap Lid Type Loose
Color Blue
Depth (Metric) Inside 500mm
Diameter (Metric) Outer 250mm
For Use With (Application) Laboratory technique, Medicinal technique
Height (Metric) 610mm
Includes Lid Handle/Two Grips
Base Material Borosilicate Glass
Capacity (Metric) 14 L
Material Aluminum