Learn More
Macherey-Nagel™ Developing Chamber
Product Code. 10640732 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
TLC Micro-Sets
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10640732 TLC Micro-Sets Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10640732 Supplier Macherey-Nagel™ Supplier No. 814021
  • ONLINE EXCLUSIVE PRICE
    493.00 NOK
    400.28 NOK / Pack of 4
    Save 92.72 NOK
    18% Off
View all promotions
Estimated Shipment: 17-11-2026
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Specifications
Quantity 4 Pk.
Type Developing Chamber
For Use With (Application) For TLC Micro-Sets
For Use With (Equipment) TLC Micro-Sets