Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
Specifications
| Light Source | UV |
| Wavelength | 400/185 nm |
| Aperture Size (Metric) | 0.6 mm |
| For Use With (Equipment) | Jasco UV970 975 1570 1575 |
| Packaging Quantity | 1 Ea. |
| Product Type | Deuterium Lamp |