Learn More
Biochrom Deuterium Lamp Replacement
Product Code. 15950591 Shop All Harvard Bioscience Products
Change view
Click to view available options
Product Type:
Replacement Lamp
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15950591 Replacement Lamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15950591 Supplier Biochrom Supplier No. 80700050
  • ONLINE EXCLUSIVE PRICE
    11210.00 NOK
    10294.47 NOK / Pack of 1
    Save 915.53 NOK
    8% Off
Estimated Shipment: 09-12-2026
Deuterium Lamp Replacement suitable for Biochrom Ultrospec 8000/9000 and Libra S80/S90.
  • For Biochrom Ultrospec 8000/9000 and Libra S80/S90
Specifications
Light Source Deuterium
Voltage 110/240 V
For Use With (Equipment) Ultrospec 8000, 9000, and Libra S80, S90 UV-Visible Spectrophotometers
Product Type Replacement Lamp