missing translation for 'onlineSavingsMsg'
Learn More

Detomidine hydrochloride, MedChemExpress

Product Code. p-200101488
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16418146

Brand: Medchem Express HYB0163A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Detomidine hydrochloride, an imidazole derivative, is a potent α2-adrenergic agonist. Detomidine hydrochloride produces dose-dependent analgesic effects.

Detomidine hydrochloride, an imidazole derivative, is a potent α2-adrenergic agonist. Detomidine hydrochloride produces dose-dependent analgesic effects.

Specifications

Chemical Name or Material Detomidine hydrochloride
Physical Form Solid
CAS 90038-01-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H15ClN2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (449.01 mM) ∣H2O : 20 mg/mL (89.80 mM; Need ultrasonic)
SMILES CC1=C(C)C(CC2=CN=CN2)=CC=C1.Cl
Molecular Weight (g/mol) 222.71
Formula Weight 222.71
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.