missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Monoclonal Antibody (OTI6H3), TrueMAB™, OriGene
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
Desmin Monoclonal antibody specifically detects Desmin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Desmin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI6H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | DES |
| Gene Accession No. | P17661, P31001, P48675 |
| Gene Alias | CMD1I; CSM1; CSM2; DES; Desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?