Learn More
Vitlab™ Desiccator with Stopcock
Product Code. 10613072
Change view
Click to view available options
Height (Metric):
190 mm
Unit Size:
Each
Product Code. Height (Metric) unitSize
10613072 190 mm Each
1 options
Product Code. 10613072
Brand: Vitlab™ 80550
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Vitlab™ Desiccator with Stopcock includes a bleed valve and an O-ring seal between the dome and the lower portion.
  • With a bleed valve and an O-ring seal between the dome and the lower portion
  • The desiccators are suitable for vacuum and are provided with a non-return valve
  • Hot crucibles should only be placed on a porcelain plate, and should not come into too close contact with the rim of the desiccator
  • The insert made of PP serves to hold the drying agent
Specifications
Material Polypropylene, Polycarbonate
Includes Stopcock
Height (Metric) 190 mm
Diameter (Metric) Inner 140 mm disc
Diameter (Metric) Outer 150 mm