Learn More
BRAND™ Desiccator with Knob Lid
Desiccator with Knob Lid includes a studded base ring for improved stability.
2375.00 NOK - 3005.00 NOK
Specifications
Material | Borosilicate Glass 3.3 |
---|---|
Product Type | Desiccator |
Includes | Knob Lid |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10728051
|
BRAND™
65031 |
150 mm |
2375.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10470673
|
BRAND™
65038 |
200 mm |
3005.00 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
- Borosilicate Glass 3.3
- DIN 12491
- Precisely ground flat flanges
- Base ring surface studded to improve stability and help prevent cracks
- Interchangeable lid
Specifications
Borosilicate Glass 3.3 | |
Desiccator | |
Knob Lid |