Learn More
asecos™ Depot PE with Pull-Down Blind - Version Plastic Rack
Product Code. p-200003660 Shop All asecos Products
Click to view available options
Depth (Metric):
1050.00 mm
1083.00 mm
Unit Size:
Each
Product Code. 17896576
Brand: asecos™ 40062
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Depot PE with Pull-Down Blind is approved for the safe storage of water pollutants and has a high resistance to oils, acids and alkalis.
  • With integrated sump pallet
  • Integrated pocket for access underneath by forklifts and forklift trucks
  • Ready for anchoring to the ground (anchor kit optional)
  • Safe storage of small and medium-sized containers
  • Rack with PE rack frames and 3 shelves
  • Dimensions shelves B x D (mm): 900 x 590 (PE)
  • Load capacity per storage level: 180 kg
Specifications
Color Grey
Capacity (English) 64.7 gal.
Capacity (Metric) 245 L
Material Polyethylene
Type Disposal Can
Length (English) 44.88 in.
Length (Metric) 1140.00 mm
Width (English) 44.88 in.
Width (Metric) 1140.00 mm
Height (English) 81.10 in.
Height (Metric) 2060.00 mm
Depth (English) 41.34 in.
Depth (Metric) 1050.00 mm
Weight (English) 264.55 lb.
Weight (Metric) 120 kg