missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic acid, MedChemExpress

Product Code. p-200100508
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16489106

Brand: Medchem Express HYN0593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Deoxycholic acid (cholanoic acid), a bile acid, is a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptorTGR5.

Deoxycholic acid (cholanoic acid), a bile acid, is a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptorTGR5.

Specifications

Chemical Name or Material Deoxycholic acid
CAS 83-44-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H40O4
Quantity 100 mg
Synonym Cholanoic Acid Desoxycholic acid
Solubility Information DMSO : ≥ 100 mg/mL (254.73 mM) ∣H2O : 1 mg/mL (2.55 mM; ultrasonic and warming and heat to 80°C)
SMILES C[C@@]1([C@@]2([H])[C@H](C)CCC(O)=O)[C@](CC2)([H])[C@@](CC[C@@]3([H])[C@@]4(CC[C@@H](O)C3)C)([H])[C@]4([H])C[C@@H]1O
Molecular Weight (g/mol) 392.57
Formula Weight 392.57
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.