Learn More
Density Standards (25 °C), Paragon Scientific™
Product Code. 13480509
Change view
Click to view available options
Density:
0.6552 g/mL (25°C)
0.7223 g/mL (25°C)
0.775 g/mL (25°C)
0.8349 g/mL (25°C)
0.8678 g/mL (25°C)
0.934 g/mL (25°C)
0.9803 g/mL (25°C)
1.0242 g/mL (25°C)
1.0657 g/mL (25°C)
1.1233 g/mL (25°C)
1.1869 g/mL (25°C)
1.2819 g/mL (25°C)
1.6143 g/mL (25°C)
Unit Size:
60mL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Density Unit Size
13480509 0.8678 g/mL (25°C) 60mL
13440509 0.6552 g/mL (25°C) 60mL
13450509 0.7223 g/mL (25°C) 60mL
13460509 0.775 g/mL (25°C) 60mL
13470509 0.8349 g/mL (25°C) 60mL
13410519 0.934 g/mL (25°C) 60mL
13400519 0.9803 g/mL (25°C) 60mL
13420519 1.0242 g/mL (25°C) 60mL
13430519 1.0657 g/mL (25°C) 60mL
13450519 1.1233 g/mL (25°C) 60mL
13440519 1.1869 g/mL (25°C) 60mL
13460519 1.2819 g/mL (25°C) 60mL
13470519 1.6143 g/mL (25°C) 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 13480509 Supplier Paragon Scientific Supplier No. DEN2505
  • ONLINE EXCLUSIVE PRICE
    1055.00 NOK
    771.49 NOK / 60mL
    Save 283.51 NOK
    26% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 25°C
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging (please refer to standard for shelf life).
  • Manufactured in the United Kingdom and available from stock for immediate dispatch
Specifications
Density 0.8678 g/mL (25°C)
Quantity 60 mL
Nominal values quoted