Learn More
Density Standards (20 °C), Paragon Scientific™
Product Code. 13430499
Change view
Click to view available options
Density:
0.6597 g/mL (20°C)
0.7264 g/mL (20°C)
0.7788 g/mL (20°C)
0.8383 g/mL (20°C)
0.8556 g/mL (20°C)
0.8708 g/mL (20°C)
0.9376 g/mL (20°C)
0.9817 g/mL (20°C)
1.0257 g/mL (20°C)
1.0677 g/mL (20°C)
1.1258 g/mL (20°C)
1.1917 g/mL (20°C)
1.2849 g/mL (20°C)
1.6226 g/mL (20°C)
Unit Size:
60mL
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Density Unit Size
13430499 0.9817 g/mL (20°C) 60mL
13480489 0.6597 g/mL (20°C) 60mL
13470489 0.7264 g/mL (20°C) 60mL
13490489 0.7788 g/mL (20°C) 60mL
13400499 0.8383 g/mL (20°C) 60mL
13420499 0.8556 g/mL (20°C) 60mL
13450499 0.8708 g/mL (20°C) 60mL
13440499 0.9376 g/mL (20°C) 60mL
13460499 1.0257 g/mL (20°C) 60mL
13490499 1.0677 g/mL (20°C) 60mL
13480499 1.1258 g/mL (20°C) 60mL
13400509 1.1917 g/mL (20°C) 60mL
13410509 1.2849 g/mL (20°C) 60mL
13420509 1.6226 g/mL (20°C) 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 13430499 Supplier Paragon Scientific Supplier No. DEN2008
  • ONLINE EXCLUSIVE PRICE
    1055.00 NOK
    771.49 NOK / 60mL
    Save 283.51 NOK
    26% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 20°C
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging with a 2 year shelf life.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • Manufactured in the United Kingdom
Specifications
Density 0.9817 g/mL (20°C)
Certifications/Compliance ISO 17025, UKAS ISO 17034
Quantity 60 mL
compliance-icons compliance-icons
Product Identifier
  • Density Standard (20 °C)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Nominal values quoted