Learn More
Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage
Product Code. 15947972 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Plates (10/Rack)
Plates (11/Rack)
Plates (12/Rack)
Plates (15/Rack)
Plates (18/Rack)
Plates (9/Rack)
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15947972 Plates (15/Rack) Each
15596480 Plates (10/Rack) Each
15920211 Plates (9/Rack) Each
15346395 Plates (18/Rack) Each
15366395 Plates (12/Rack) Each
15549831 Plates (11/Rack) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15947972 Supplier Thermo Scientific™ Supplier No. 1950832
A convenient, streamlined process to maximize sample storage. Our freezer racking and tubing solutions are designed to hold tubes ranging from 200μL to 5.0μL.
A convenient, streamlined process to maximize sample storage in your laboratory. Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage are designed to hold tubes ranging from 200μL up to 5.0μL, offering maximized sample storage capacity for ultra-low chest freezers and large-capacity cryogenic freezers.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Description Racks for Tubes (cryogenic and ultra-low chest freezers)
Color Silver
Holds Plates (15/Rack)
Shape Rectangular
For Use With (Equipment) Matrix 200μL Screwtop universal tubes, 500μL storage tubes, 500μL storage tubes with DuraSeal, 0.5mL Nunc Cryobank tubes
Freezer Compatibility (Capacity) Thermo Scientific CryoPlus 1 (hold 7 racks), CryoPlus 2 (holds 16 racks), CryoPlus 3 (holds 30 racks), CryoPlus 4 (holds 51 racks)
Thermo Scientific mechanical cryogenic chest freezer 6.8 cu. ft. (holds 18 racks), 10.3 cu. ft. (holds 27 racks)
Width (English) 5.3 in.
Height (English) 22.5 in.
Depth (English) 3.6 in.
Depth (Metric) 9.3 cm
Product Type Tube Rack (Cryogenic and Ultra-Low Chest Freezers)
Height (Metric) 57.2 cm
Width (Metric) 13.5 cm