missing translation for 'onlineSavingsMsg'
Learn More

Demecolcine, ≥98%, MP Biomedicals™

Product Code. 11491621 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491621 5 mg 5mg
11481621 1 mg 1mg
11401631 10 mg 10mg
11411631 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11491621 Supplier MP Biomedicals Supplier No. 0219019505
  • ONLINE EXCLUSIVE PRICE
    2760.00 NOK
    2625.65 NOK / 5mg
    Save 134.35 NOK
    4% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 477-30-5
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00075459
InChI Key NNJPGOLRFBJNIW-UHFFFAOYNA-N
Synonym demecolcine, colcemid, colchamine, --demecolcine, reichstein's f, colcemide, demecolcin, desmecolcine, santavy's substance f, --colchamine
PubChem CID 220401
ChEBI CHEBI:4393
SMILES CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12

Specifications

Color Yellow
Assay Percent Range >98%
Quantity 5 mg
IUPAC Name 3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Formula Weight 371.4
Physical Form Powder
Chemical Name or Material Demecolcine, Antineoplastic agent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.