Learn More
Lenz Laborglasintrumente Delivery Adapters Straight with Suction Tube
Product Code. 10599741
Change view
Click to view available options
Standard Taper Joint No.:
NS 14/23
NS 19/26
NS 29/32
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No. Unit Size
10599741 NS 29/32 Each
10131811 NS 14/23 Each
10589741 NS 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10599741 Supplier Lenz Laborglasintrumente Supplier No. 05266004
  • ONLINE EXCLUSIVE PRICE
    339.00 NOK
    283.68 NOK / Pack of 1
    Save 55.32 NOK
    16% Off
Estimated Shipment: 29-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Delivery adapters
  • Straight
  • With suction tube
  • With NS socket and NS cone
  • With drain tip
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Standard Taper Joint No. NS 29/32