Learn More
- Home
- Products
- Water Purification
- Water Purification Filters and Cartridges
- Thermo Scientific™ Deionizer Cartridges
Thermo Scientific™ Deionizer Cartridges
Product Code. 11799428 Shop All Thermo Scientific Products| Product Code. | For Use With (Equipment) | Resin Type | For Use With (Application) | unitSize | Capacity |
|---|---|---|---|---|---|
| 11799428 | Milli-Q Millipore Water Systems | Mixed Bed | Ion Exchange, Ultrapure | Each | - |
| 12740666 | Bantam™ Deionizer, B-Pure™ | Strong Anion Exchange | Anion Removal | Each | 1680 grains |
| 14871651 | Nanopure | Mixed Bed | Ultrapure | Each | - |
| 15342178 | B-Pure™ | Activated Carbon | Organic Removal | 1 unit | 5000 gal. (as CaCO3) |
| 15352178 | B-Pure™ | Mixed Bed | Ultrapure | 1 unit | - |
| 15362178 | B-Pure™ | Oxygen Removal Resin | Oxygen Removal | 1 unit | - |
| 12331683 | B-Pure™ | Ultrapure, Oxygen Removal | Ultrapure, Oxygen and Organic Removal | Each | - |
| 15372178 | B-Pure™ | Mixed Bed, Ultrapure, Oxygen and Organic Removal | Ultrapure and Oxygen Removal Resin | 1 unit | - |
| 11789428 | Milli-Q Millipore Water Systems | Super-C Carbon | Organic Removal | Each | - |
| 10071893 | Bantam™ Deionizer, B-Pure™ | Mixed Bed | - | Each | 875 grains |
| 10589690 | B-Pure™ | Porous Strong Ion Exchange | Oxygen Removal | Each | 30 g |
| 10041953 | Bantam™ Deionizer, B-Pure™ | Activated Carbon | Organic Removal | Each | 5000 gal. |
| 12311278 | Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | Cation Removal | Each | 3170 grains |
| 11746189 | Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | Ultrapure and Organic Removal | Each | 785 grains/1000 gal. |
| 10451820 | B-Pure™ | Macroreticular, Activated Carbon | Pretreatment | Each | 5000 gal. |
| 10297093 | B-Pure™ | Macroreticular, Activated Carbon | Pretreatment Macropure | Each | 2000 gal. |
| 10074067 | B-Pure™ | Mixed Bed | Ultrapure | Each | 550 grains |
| 15312168 | B-Pure™ | Porous Strong Anion Exchange | Oxygen Removal | 1 unit | 18 g |
| 15322168 | B-Pure™ | Activated Carbon | Organic Removal | 1 unit | 5000 gal. |
| 15297643 | B-Pure™ | Oxygen Removal Mixed Bed | Oxygen and demineralization | Each | 275 grains/9 g |
| 10125767 | B-Pure™ | Mixed Bed, Activated Carbon | Ultrapure and Organic Removal | Each | 275 grains/2000 gal. |
| 11706289 | B-Pure™ | Extruded Carbon | Organic Removal | Each | 5000 gal. |
| 12730746 | B-Pure™ | Ultrapure, Oxygen Removal | Ultrapure and Organic Removal | Each | 455 grains/30 g |
| 10081893 | B-Pure™ | Ultrapure, Oxygen and Organic Removal | Ultrapure, Oxygen and Organic Removal | Each | 365 grains/2000 gal./12 g |
| 10501673 | Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated | Two Bed Ion Exchange | High capacity, two-bed ion exchange | Each | 1760 grains |
Product Code. 11799428
Brand: Thermo Scientific™ D8832
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
Specifications
| Description | MQ Cartridge Ion Exchange, Ultrapure Mixed-bed |
| Purification Method | Ion Exchange |
| Resin Type | Mixed Bed |
| For Use With (Application) | Ion Exchange, Ultrapure |
| For Use With (Equipment) | Milli-Q Millipore Water Systems |
| Type | Deionizer Cartridge |