Learn More
- Home
- Products
- Water Purification
- Water Purification Filters and Cartridges
- Thermo Scientific™ Deionizer Cartridges
Thermo Scientific™ Deionizer Cartridges
Product Code. 11789428 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Milli-Q Millipore Water Systems
Nanopure
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Mixed Bed, Ultrapure, Oxygen and Organic Removal
Oxygen Removal Mixed Bed
Oxygen Removal Resin
Porous Strong Anion Exchange
Porous Strong Ion Exchange
Strong Anion Exchange
Strong Cation Exchange
Super-C Carbon
Two Bed Ion Exchange
Ultrapure, Oxygen Removal
Ultrapure, Oxygen and Organic Removal
For Use With (Application):
Anion Removal
Cation Removal
High capacity, two-bed ion exchange
Ion Exchange, Ultrapure
Organic Removal
Oxygen Removal
Oxygen and demineralization
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure and Oxygen Removal Resin
Ultrapure, Oxygen and Organic Removal
Unit Size:
1 unit
Each
Capacity:
1680 grains
1760 grains
18 g
2000 gal.
275 grains/2000 gal.
275 grains/9 g
30 g
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
5000 gal.
5000 gal. (as CaCO3)
550 grains
785 grains/1000 gal.
875 grains
| Product Code. | For Use With (Equipment) | Resin Type | For Use With (Application) | unitSize | Capacity |
|---|---|---|---|---|---|
| 11789428 | Milli-Q Millipore Water Systems | Super-C Carbon | Organic Removal | Each | - |
| 12740666 | Bantam™ Deionizer, B-Pure™ | Strong Anion Exchange | Anion Removal | Each | 1680 grains |
| 14871651 | Nanopure | Mixed Bed | Ultrapure | Each | - |
| 15342178 | B-Pure™ | Activated Carbon | Organic Removal | 1 unit | 5000 gal. (as CaCO3) |
| 15352178 | B-Pure™ | Mixed Bed | Ultrapure | 1 unit | - |
| 15362178 | B-Pure™ | Oxygen Removal Resin | Oxygen Removal | 1 unit | - |
| 12331683 | B-Pure™ | Ultrapure, Oxygen Removal | Ultrapure, Oxygen and Organic Removal | Each | - |
| 15372178 | B-Pure™ | Mixed Bed, Ultrapure, Oxygen and Organic Removal | Ultrapure and Oxygen Removal Resin | 1 unit | - |
| 11799428 | Milli-Q Millipore Water Systems | Mixed Bed | Ion Exchange, Ultrapure | Each | - |
| 10071893 | Bantam™ Deionizer, B-Pure™ | Mixed Bed | - | Each | 875 grains |
| 10589690 | B-Pure™ | Porous Strong Ion Exchange | Oxygen Removal | Each | 30 g |
| 10041953 | Bantam™ Deionizer, B-Pure™ | Activated Carbon | Organic Removal | Each | 5000 gal. |
| 12311278 | Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | Cation Removal | Each | 3170 grains |
| 11746189 | Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | Ultrapure and Organic Removal | Each | 785 grains/1000 gal. |
| 10451820 | B-Pure™ | Macroreticular, Activated Carbon | Pretreatment | Each | 5000 gal. |
| 10297093 | B-Pure™ | Macroreticular, Activated Carbon | Pretreatment Macropure | Each | 2000 gal. |
| 10074067 | B-Pure™ | Mixed Bed | Ultrapure | Each | 550 grains |
| 15312168 | B-Pure™ | Porous Strong Anion Exchange | Oxygen Removal | 1 unit | 18 g |
| 15322168 | B-Pure™ | Activated Carbon | Organic Removal | 1 unit | 5000 gal. |
| 15297643 | B-Pure™ | Oxygen Removal Mixed Bed | Oxygen and demineralization | Each | 275 grains/9 g |
| 10125767 | B-Pure™ | Mixed Bed, Activated Carbon | Ultrapure and Organic Removal | Each | 275 grains/2000 gal. |
| 11706289 | B-Pure™ | Extruded Carbon | Organic Removal | Each | 5000 gal. |
| 12730746 | B-Pure™ | Ultrapure, Oxygen Removal | Ultrapure and Organic Removal | Each | 455 grains/30 g |
| 10081893 | B-Pure™ | Ultrapure, Oxygen and Organic Removal | Ultrapure, Oxygen and Organic Removal | Each | 365 grains/2000 gal./12 g |
| 10501673 | Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated | Two Bed Ion Exchange | High capacity, two-bed ion exchange | Each | 1760 grains |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
Specifications
| Description | MQ Cartridge Organic Removal, Super-C Carbon |
| Resin Type | Super-C Carbon |
| For Use With (Application) | Organic Removal |
| For Use With (Equipment) | Milli-Q Millipore Water Systems |
| Type | Deionizer Cartridge |