Learn More
Deglucopyranosyl Ethoxy Etoposide, TRC
Specifications
Specifications
| Chemical Name or Material | 4'-O-Demethyl-1-O-ethylepipodophyllotoxin |
| CAS | 102306-95-6 |
| Molecular Formula | C23 H24 O8 |
| Quantity | 50 mg |
| Synonym | Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-ethoxy-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, [5R-(5α,5aβ,8aα,9β)]- (9CI), (5R,5aR,8aR,9S)-9-Ethoxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydro[2]benzofuro[5,6-f][1,3]benzod |
| InChI Formula | InChI=1S/C23H24O8/c1-4-28-22-13-8-16-15(30-10-31-16)7-12(13)19(20-14(22)9-29-23(20)25)11-5-17(26-2)21(24)18(6-11)27-3/h5-8,14,19-20,22,24H,4,9-10H2,1-3H3/t14-,19+,20-,22+/m0/s1 |
| SMILES | CCO[C@H]1[C@H]2COC(=O)[C@@H]2[C@H](c3cc(OC)c(O)c(OC)c3)c4cc5OCOc5cc14 |
| IUPAC Name | (5S,5aR,8aR,9R)-5-ethoxy-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one |
| Molecular Weight (g/mol) | 428.432 |
| Formula Weight | 428.147 |